Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FXYD1
Entrez Gene ID
476487
Gene Name
FXYD domain containing ion transport regulator 1
Gene Aliases
PLM
Chromosome Location
Chr.1: 117436239 - 117440619 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195142.1 NP_001182071.1 4 - 5 70
NM_001195143.1 NP_001182072.1 4 - 5 70
XM_005615754.3 XP_005615811.2 4 - 5 70
GenBank mRNA M63934.1 - 4 - 5 268 70

View Full Product Details