Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRP54
Entrez Gene ID
403953
Gene Name
signal recognition particle 54
Gene Aliases
-
Chromosome Location
Chr.8: 13800275 - 13837799 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003272.1 NP_001003272.1 6 - 7 732 70
XM_005623180.3 XP_005623237.1 5 - 6 932 70
XM_005623181.3 XP_005623238.1 5 - 6 538 70
GenBank mRNA X16318.1 - - 732 70

View Full Product Details