Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB8A
Entrez Gene ID
403773
Gene Name
RAB8A, member RAS oncogene family
Gene Aliases
RAB8
Chromosome Location
Chr.20: 46334601 - 46351907 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003152.1 NP_001003152.1 7 - 8 541 84
GenBank mRNA X56385.1 - - 541 84

View Full Product Details