Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF9
Entrez Gene ID
477510
Gene Name
serine and arginine rich splicing factor 9
Gene Aliases
SFRS9
Chromosome Location
Chr.26: 16363275 - 16369699 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534706.6 XP_534706.1 3 - 4 612 75

View Full Product Details