Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MZB1
Entrez Gene ID
100688475
Gene Name
marginal zone B and B1 cell specific protein
Gene Aliases
-
Chromosome Location
Chr.2: 34710487 - 34712660 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003433755.4 XP_003433803.2 - 361 93

View Full Product Details