Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
474753
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
-
Chromosome Location
Chr.11: 51587728 - 51596911 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005626745.3 XP_005626802.1 3 - 4 727 86
XM_860937.5 XP_866030.2 3 - 4 730 86

View Full Product Details