Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPIL1
Entrez Gene ID
474891
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
-
Chromosome Location
Chr.12: 5916524 - 5936276 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014118403.2 XP_013973878.1 2 - 3 579 95

View Full Product Details