Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH4
Entrez Gene ID
480116
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Gene Aliases
-
Chromosome Location
Chr.7: 40010412 - 40022425 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005622974.2 XP_005623031.2 1 - 2 158 76

View Full Product Details