Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GM2A
Entrez Gene ID
100855818
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.4: 58002438 - 58016003 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639011.4 XP_003639059.1 3 - 4 707 77

View Full Product Details