Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMM22
Entrez Gene ID
480639
Gene Name
translocase of inner mitochondrial membrane 22
Gene Aliases
-
Chromosome Location
Chr.9: 44823368 - 44830375 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537758.6 XP_537758.1 2 - 3 766 70

View Full Product Details