Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP3
Entrez Gene ID
480505
Gene Name
dual specificity phosphatase 3
Gene Aliases
-
Chromosome Location
Chr.9: 19518302 - 19531000 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116479.1 XP_013971954.1 - 454 94

View Full Product Details