Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD15
Entrez Gene ID
480401
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
-
Chromosome Location
Chr.8: 50182282 - 50193262 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537522.6 XP_537522.1 1 - 2 1754 74

View Full Product Details