Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS25
Entrez Gene ID
479412
Gene Name
ribosomal protein S25
Gene Aliases
-
Chromosome Location
Chr.5: 14833804 - 14836376 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536549.5 XP_536549.3 1 - 1 161 60

View Full Product Details