Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED1
Entrez Gene ID
484948
Gene Name
transmembrane p24 trafficking protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 50395862 - 50399118 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_542064.6 XP_542064.1 1 - 2 331 80

View Full Product Details