Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH14
Entrez Gene ID
100856223
Gene Name
retinol dehydrogenase 14
Gene Aliases
-
Chromosome Location
Chr.17: 13843278 - 13848498 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639576.4 XP_003639624.3 1 - 2 470 72

View Full Product Details