Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IER3
Entrez Gene ID
481708
Gene Name
immediate early response 3
Gene Aliases
-
Chromosome Location
Chr.12: 529002 - 530328 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_538829.6 XP_538829.1 1 - 2 315 67

View Full Product Details