Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YRDC
Entrez Gene ID
100855794
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Gene Aliases
-
Chromosome Location
Chr.15: 4724468 - 4728659 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639501.3 XP_003639549.1 2 - 3 531 87

View Full Product Details