Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISG20
Entrez Gene ID
488729
Gene Name
interferon stimulated exonuclease gene 20
Gene Aliases
-
Chromosome Location
Chr.3: 51822642 - 51833665 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_545847.6 XP_545847.2 3 - 4 731 75

View Full Product Details