Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGT1
Entrez Gene ID
486400
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.26: 28301455 - 28315029 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005636554.3 XP_005636611.1 4 - 5 662 97

View Full Product Details