Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YARS2
Entrez Gene ID
486614
Gene Name
tyrosyl-tRNA synthetase 2
Gene Aliases
-
Chromosome Location
Chr.27: 16139462 - 16162701 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_543740.6 XP_543740.1 3 - 4 1146 76

View Full Product Details