Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBMX2
Entrez Gene ID
492135
Gene Name
RNA binding motif protein X-linked 2
Gene Aliases
-
Chromosome Location
Chr.X: 101533732 - 101547147 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641806.3 XP_005641863.1 4 - 5 429 88

View Full Product Details