Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
491271
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
-
Chromosome Location
Chr.9: 49827584 - 49834872 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116953.2 XP_013972428.1 4 - 5 873 59

View Full Product Details