Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL18
Entrez Gene ID
489881
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
-
Chromosome Location
Chr.6: 12434983 - 12479702 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_546999.6 XP_546999.3 - 2118 87

View Full Product Details