Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTA
Entrez Gene ID
608826
Gene Name
T cell leukemia translocation altered
Gene Aliases
-
Chromosome Location
Chr.20: 39820845 - 39824526 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845960.5 XP_851053.1 1 - 2 601 62

View Full Product Details