Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LEAP2
Entrez Gene ID
609750
Gene Name
liver enriched antimicrobial peptide 2
Gene Aliases
-
Chromosome Location
Chr.11: 21156051 - 21157197 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847079.5 XP_852172.1 2 - 3 420 64

View Full Product Details