Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS2
Entrez Gene ID
608318
Gene Name
mitochondrial ribosomal protein S2
Gene Aliases
-
Chromosome Location
Chr.9: 51211701 - 51214465 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_845298.4 XP_850391.1 2 - 3 251 93

View Full Product Details