Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC42EP5
Entrez Gene ID
611420
Gene Name
CDC42 effector protein 5
Gene Aliases
-
Chromosome Location
Chr.1: 102838163 - 102842586 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849090.5 XP_854183.1 1 - 2 278 152

View Full Product Details