Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRP9
Entrez Gene ID
611923
Gene Name
signal recognition particle 9
Gene Aliases
-
Chromosome Location
Chr.7: 39012142 - 39023009 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849646.5 XP_854739.1 1 - 2 197 79

View Full Product Details