Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YPEL2
Entrez Gene ID
610066
Gene Name
yippee like 2
Gene Aliases
-
Chromosome Location
Chr.9: 33872730 - 33935877 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847459.5 XP_852552.1 1 - 2 158 102

View Full Product Details