Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO46
Entrez Gene ID
612248
Gene Name
F-box protein 46
Gene Aliases
-
Chromosome Location
Chr.1: 109863907 - 109878908 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849986.4 XP_855079.2 1 - 2 108 88

View Full Product Details