Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53B
Entrez Gene ID
611311
Gene Name
family with sequence similarity 53 member B
Gene Aliases
-
Chromosome Location
Chr.28: 33961810 - 34077075 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108922.2 XP_013964397.1 3 - 4 453 92
XM_014108923.2 XP_013964398.1 3 - 4 468 92
XM_022411802.1 XP_022267510.1 4 - 5 625 92
XM_022411803.1 XP_022267511.1 3 - 4 422 92
XM_022411804.1 XP_022267512.1 2 - 3 317 92
XM_848965.5 XP_854058.3 3 - 4 567 92

View Full Product Details