Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC24
Entrez Gene ID
610507
Gene Name
zinc finger CCHC-type containing 24
Gene Aliases
-
Chromosome Location
Chr.4: 29103393 - 29161626 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_848028.6 XP_853121.1 2 - 3 541 89

View Full Product Details