Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCD
Entrez Gene ID
611081
Gene Name
IQ motif containing D
Gene Aliases
-
Chromosome Location
Chr.26: 10575116 - 10602139 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022410799.1 XP_022266507.1 - 1325 84

View Full Product Details