Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSM6
Entrez Gene ID
612329
Gene Name
LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.15: 44651830 - 44666418 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003432044.4 XP_003432092.1 4 - 5 464 118
XM_850069.4 XP_855162.2 3 - 4 379 118

View Full Product Details