Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD5
Entrez Gene ID
607891
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
-
Chromosome Location
Chr.27: 1941446 - 1943787 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_844743.5 XP_849836.2 2 - 2 373 134

View Full Product Details