Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XKRX
Entrez Gene ID
609590
Gene Name
XK related X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 74854947 - 74872918 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641559.3 XP_005641616.1 3 - 3 1883 81

View Full Product Details