Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PITHD1
Entrez Gene ID
612304
Gene Name
PITH domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 75714592 - 75721301 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022413403.1 XP_022269111.1 4 - 5 832 141
XM_850047.5 XP_855140.2 5 - 6 547 141

View Full Product Details