Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFIT5
Entrez Gene ID
486788
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
-
Chromosome Location
Chr.28: 4318616 - 4329811 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005637512.3 XP_005637569.1 3 - 3 839 149
XM_005637513.3 XP_005637570.1 2 - 2 751 149

View Full Product Details