Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4D5
Entrez Gene ID
479404
Gene Name
olfactory receptor family 4 subfamily D member 5
Gene Aliases
cOR4D5
Chromosome Location
Chr.5: 10546265 - 10547209 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536541.3 XP_536541.3 - 628 141

View Full Product Details