Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAICS
Entrez Gene ID
100855769
Gene Name
phosphoribosylaminoimidazole carboxylase and phosphoribosylaminoimidazolesuccinocarboxamide synthase
Gene Aliases
KIAA1211
Chromosome Location
Chr.13: 48296232 - 48682650 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003639458.4 XP_003639506.1 5 - 6 737 128
XM_022427005.1 XP_022282713.1 18 - 19 5638 128
XM_022427006.1 XP_022282714.1 17 - 18 5530 128
XM_022427008.1 XP_022282716.1 21 - 22 6031 128
XM_022427009.1 XP_022282717.1 18 - 19 5540 128
XM_022427010.1 XP_022282718.1 17 - 18 5323 128
XM_022427011.1 XP_022282719.1 20 - 21 5895 128
XM_022427012.1 XP_022282720.1 16 - 17 5257 128
XM_022427013.1 XP_022282721.1 19 - 20 5730 128
XM_022427015.1 XP_022282723.1 11 - 12 2045 128
XM_022427016.1 XP_022282724.1 11 - 12 1947 128

View Full Product Details