Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB4
Entrez Gene ID
489474
Gene Name
zinc finger and BTB domain containing 4
Gene Aliases
-
Chromosome Location
Chr.5: 32384353 - 32401658 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205162.1 NP_001192091.1 3 - 3 1332 116
XM_005619519.3 XP_005619576.1 6 - 6 1668 116
XM_005619522.2 XP_005619579.1 4 - 4 1465 116
XM_005619523.3 XP_005619580.1 4 - 4 1520 116
XM_005619524.3 XP_005619581.1 4 - 4 1435 116
XM_014113199.2 XP_013968674.1 8 - 8 1790 116
XM_022418031.1 XP_022273739.1 6 - 6 1660 116
XM_022418032.1 XP_022273740.1 5 - 5 1576 116

View Full Product Details