Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRK
Entrez Gene ID
100684309
Gene Name
CRK proto-oncogene, adaptor protein
Gene Aliases
-
Chromosome Location
Chr.9: 45594657 - 45627499 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003435202.4 XP_003435250.1 2 - 3 925 140

View Full Product Details