Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD1
Entrez Gene ID
489984
Gene Name
THUMP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 24607053 - 24615916 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_547104.5 XP_547104.2 3 - 4 724 186

View Full Product Details