Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ybx1
Entrez Gene ID
100769813
Gene Name
Y box binding protein 1
Gene Aliases
I79_022277
Chromosome Location
Chr.Un|NW_003615864: 43827 - 59571 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653625.1 XP_007651815.1 6 - 7 905 116

View Full Product Details