Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa9
Entrez Gene ID
100689405
Gene Name
heat shock 70kDa protein 9 (mortalin)
Gene Aliases
GRP-75, I79_009139
Chromosome Location
Chr.Un|NW_003613902: 467258 - 487928 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246829.1 NP_001233758.1 7 - 8 969 81
GenBank mRNA JI875975.1 - 7 - 8 844 81
JP065007.1 - 7 - 8 844 81
U92313.1 - 7 - 8 969 81

View Full Product Details