Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcb1b
Entrez Gene ID
100682537
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 1B
Gene Aliases
PGP2, PGY2
Chromosome Location
Chr.Un|NW_003614224: 748705 - 819221 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243989.1 NP_001230918.1 19 - 20 2482 68
XM_007646774.1 XP_007644964.1 20 - 21 2646 68
XM_007646775.1 XP_007644965.1 18 - 19 2421 68
XM_007646776.1 XP_007644966.1 20 - 21 2646 68
GenBank mRNA M17896.1 - 6 - 7 611 68
M60041.1 - 19 - 20 2482 68

View Full Product Details