Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cstf1
Entrez Gene ID
100761066
Gene Name
cleavage stimulation factor, 3' pre-RNA, subunit 1, 50kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613850: 1162851 - 1171799 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501440.2 XP_003501488.1 2 - 3 283 70
XM_007642098.1 XP_007640288.1 2 - 3 339 70
GenBank mRNA JI863144.1 - 2 - 3 272 70
JP056671.1 - 2 - 3 272 70

View Full Product Details