Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sc5d
Entrez Gene ID
100752960
Gene Name
sterol-C5-desaturase
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615094: 108605 - 121568 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511371.2 XP_003511419.1 3 - 4 486 107
XM_007651891.1 XP_007650081.1 3 - 4 389 107
GenBank mRNA JP054188.1 - 3 - 4 488 107

View Full Product Details