The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_007649613.1 | XP_007647803.1 | 5 - 6 | 799 | 77 |
XM_007649614.1 | XP_007647804.1 | 5 - 6 | 795 | 77 | |
XM_007649615.1 | XP_007647805.1 | 5 - 6 | 630 | 77 | |
XM_007649616.1 | XP_007647806.1 | 4 - 5 | 590 | 77 | |
XM_007649617.1 | XP_007647807.1 | 4 - 5 | 586 | 77 | |
XM_007649618.1 | XP_007647808.1 | 5 - 6 | 749 | 77 | |
XM_007649619.1 | XP_007647809.1 | 4 - 5 | 540 | 77 |