Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prickle4
Entrez Gene ID
100772474
Gene Name
prickle homolog 4 (Drosophila)
Gene Aliases
I79_017053
Chromosome Location
Chr.Un|NW_003614620: 429400 - 435996 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649613.1 XP_007647803.1 6 - 7 993 58
XM_007649614.1 XP_007647804.1 6 - 7 989 58
XM_007649615.1 XP_007647805.1 6 - 7 824 58
XM_007649616.1 XP_007647806.1 5 - 6 784 58
XM_007649617.1 XP_007647807.1 5 - 6 780 58
XM_007649618.1 XP_007647808.1 6 - 7 943 58
XM_007649619.1 XP_007647809.1 5 - 6 734 58

View Full Product Details